Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163358.1 | NP_001156830.1 | 6 | 692 | ||
NM_178247.3 | NP_839978.1 | 7 | 778 | |||
GenBank mRNA | AF490345.1 | - | 7 | 764 | ||
AK145642.1 | - | 7 | 779 | |||
AK166752.1 | - | 7 | 776 |