Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144829.2 | NP_001138301.1 | 2 | 483 | ||
NM_001144830.2 | NP_001138302.1 | 1 | 693 | |||
NM_001461.3 | NP_001452.2 | 2 | 473 | |||
XM_005272946.4 | XP_005273003.1 | 2 | 447 | |||
XM_005272947.4 | XP_005273004.1 | 2 | 191 | |||
XM_005272948.4 | XP_005273005.1 | 2 | 174 | |||
XM_006711244.3 | XP_006711307.1 | 2 | 463 | |||
XM_006711245.3 | XP_006711308.1 | 2 | 463 | |||
XM_011509350.2 | XP_011507652.1 | 2 | 478 | |||
XM_011509351.2 | XP_011507653.1 | 2 | 399 | |||
XM_017000801.1 | XP_016856290.1 | 2 | 411 | |||
XM_017000802.1 | XP_016856291.1 | 2 | 181 | |||
GenBank mRNA | AK022172.1 | - | 1 | 683 | ||
AK222728.1 | - | 2 | 188 | |||
AK225739.1 | - | 2 | 198 | |||
AK314647.1 | - | 2 | 401 | |||
BC035687.1 | - | 1 | 366 | 5751978 | ||
DA354058.1 | - | 2 | 475 | |||
DB230162.1 | - | 2 | 463 | |||
L37080.1 | - | 2 | 159 | |||
Z47553.1 | - | 2 | 165 |