Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178418.4 | NP_848505.2 | 5 | 745 | ||
GenBank mRNA | AK133178.1 | - | 5 | 731 | ||
BC050811.1 | - | 5 | 747 | 6772771 |