Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Imp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178601.3 | NP_848716.2 | 9 | 2092 | ||
GenBank mRNA | AK013334.1 | - | 9 | 2021 | ||
AK084933.1 | - | 9 | 2028 |