Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178605.4 | NP_848720.1 | 5 | 1234 | ||
GenBank mRNA | AK013251.1 | - | 5 | 1235 |