Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178652.2 | NP_848767.1 | 5 | 603 | ||
GenBank mRNA | AK050321.1 | - | 5 | 450 | ||
AK075916.1 | - | 5 | 603 | |||
AK087935.1 | - | 5 | 448 | |||
BC032054.1 | - | 194 | ||||
BC051540.1 | - | 4 | 333 | |||
BC096051.1 | - | 5 | 387 |