Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178654.4 | NP_848769.2 | 2 | 501 | ||
GenBank mRNA | AK028469.1 | - | 2 | 488 | ||
AK046248.1 | - | 2 | 418 | |||
AK049713.1 | - | 2 | 488 | |||
AK077910.1 | - | 2 | 481 | |||
AK133298.1 | - | 2 | 490 | |||
AK165363.1 | - | 1 | 2130 | |||
AK171092.1 | - | 2 | 432 | |||
BC052073.1 | - | 3 | 424 | 6400440 |