Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pirt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178656.3 | NP_848771.1 | 2 | 542 | ||
GenBank mRNA | AK041182.1 | - | 2 | 543 | ||
AK051158.1 | - | 2 | 488 | |||
AK051285.1 | - | 2 | 552 | |||
AK051645.1 | - | 2 | 543 | |||
AK084056.1 | - | 2 | 507 | |||
BC147296.1 | - | 2 | 440 | 9056340 | ||
BC147297.1 | - | 2 | 440 | 9056338 | ||
EU447173.1 | - | 2 | 533 |