Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270792.1 | NP_001257721.1 | 3 | 490 | ||
NM_001270793.1 | NP_001257722.1 | 3 | 490 | |||
GenBank mRNA | AK087852.1 | - | 3 | 468 | ||
AK135749.1 | - | 3 | 468 | |||
AK139641.1 | - | 3 | 467 | |||
BC086484.1 | - | 3 | 463 | 30476881 |
Gene Symbol: Gm10436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105254.1 | NP_001098724.1 | 3 | 542 | ||
GenBank mRNA | AK139576.1 | - | 2 | 127 | ||
BC094894.1 | - | 2 | 104 |
Gene Symbol: Oog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178657.5 | NP_848772.3 | 3 | 490 | ||
GenBank mRNA | AB050008.3 | - | 1 | 80 | ||
AB077322.2 | - | 1 | 80 | |||
AK143386.1 | - | 3 | 468 | |||
AK145346.1 | - | 3 | 470 | |||
CF915726.1 | - | 3 | 490 |