Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178680.4 | NP_848795.3 | 12, 13 | 1804 | ||
GenBank mRNA | AK052294.1 | - | 12, 13 | 1731 | ||
AK081821.1 | - | 12, 13 | 1743 | |||
AK084418.1 | - | 12, 13 | 1772 | |||
AK168968.1 | - | 12, 13 | 1773 | |||
AK169274.1 | - | 12, 13 | 1743 | |||
BC084585.1 | - | 12, 13 | 1737 | 30429260 |