Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016231.4 | NP_057315.3 | 11 | 1876 | ||
XM_005257988.2 | XP_005258045.1 | 10 | 1804 | |||
GenBank mRNA | AF180819.1 | - | 11 | 1874 | ||
AF197898.1 | - | 11 | 1876 | |||
AL161970.1 | - | 3 | 326 | |||
BC064663.1 | - | 11 | 2388 | 6527673 | ||
DA573077.1 | - | 1 | 56 |