Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178698.5 | NP_848813.3 | 2, 3 | 460 | ||
GenBank mRNA | AK046527.1 | - | 2, 3 | 421 | ||
AK047322.1 | - | 2, 3 | 258 | |||
AK052446.1 | - | 2, 3 | 434 | |||
AK080858.1 | - | 2, 3 | 414 | |||
BC055060.1 | - | 2, 3 | 410 | 6528864 |