Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178700.5 | NP_848815.2 | 1, 2 | 422 | ||
GenBank mRNA | AK044889.1 | - | 1, 2 | 423 | ||
AK045986.1 | - | 1, 2 | 376 | |||
AK077782.1 | - | 1, 2 | 423 |