Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178717.3 | NP_848832.1 | 1 | 1148 | ||
GenBank mRNA | AK043414.1 | - | 1 | 1173 | ||
AK046367.1 | - | 1 | 899 | |||
AK084001.1 | - | 1 | 1150 | |||
AY170825.1 | - | 1 | 464 | |||
AY633762.1 | - | 1 | 464 | |||
BC053073.1 | - | 1 | 570 | 5686363 |
Set Membership: |
G-protein coupled receptor |