Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gskip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178613.3 | NP_848728.2 | 3 | 2766 | ||
GenBank mRNA | AK029484.1 | - | 4 | 2619 |