Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 2 | 250 | ||
GenBank mRNA | AK003990.1 | - | 2 | 206 | ||
AK039949.1 | - | 2 | 250 | |||
AK041783.1 | - | 244 | ||||
AK084395.1 | - | 2 | 244 | |||
AK086633.1 | - | 242 | ||||
AK144975.1 | - | 2 | 251 | |||
AK159740.1 | - | 2 | 262 | |||
AK167806.1 | - | 2 | 244 | |||
AK168510.1 | - | 2 | 206 | |||
AK169066.1 | - | 2 | 240 | |||
AK169351.1 | - | 2 | 207 | |||
BC119178.1 | - | 2 | 244 | 8733927 | ||
BC119180.1 | - | 2 | 244 | 8733929 |