Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178618.3 | NP_848733.2 | 1, 2 | 654 | ||
GenBank mRNA | AK028566.1 | - | 1, 2 | 601 | ||
BC085279.1 | - | 1, 2 | 615 | 30438273 |