Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178624.6 | NP_848739.1 | 9 | 652 | ||
GenBank mRNA | AK013142.1 | - | 10 | 786 | ||
AK039010.1 | - | 9 | 652 | |||
AK044693.1 | - | 9 | 649 | |||
AK045742.1 | - | 9 | 639 | |||
AK089994.1 | - | 9 | 628 | |||
AK141177.1 | - | 9 | 642 | |||
AK163472.1 | - | 9 | 648 | |||
BC051530.1 | - | 5 | 466 | |||
BC096582.1 | - | 9 | 638 | 3963599 | ||
BC145665.1 | - | 9 | 632 | 40130968 | ||
BC145998.1 | - | 9 | 632 | 40131301 |