Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178628.5 | NP_848743.1 | 3 | 597 | ||
GenBank mRNA | AK046919.1 | - | 2 | 327 | ||
AK050339.1 | - | 3 | 434 | |||
AK140185.1 | - | 2 | 341 | |||
AK147311.1 | - | 3 | 1089 | |||
BC050784.1 | - | 2 | 326 | 6595181 |