Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ispd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289502.1 | NP_001276431.1 | 2 | 599 | ||
NM_001289503.1 | NP_001276432.1 | 2 | 181 | |||
NM_001289504.1 | NP_001276433.1 | 2 | 599 | |||
NM_178629.6 | NP_848744.2 | 2 | 599 | |||
GenBank mRNA | AK016317.1 | - | 2 | 149 | ||
AK039004.1 | - | 2 | 467 | |||
AK043122.1 | - | 2 | 182 | |||
AK045940.1 | - | 2 | 601 | |||
AK051322.1 | - | 2 | 292 | |||
BC050993.1 | - | 2 | 437 | |||
BC086665.1 | - | 2 | 530 | 30612014 | ||
BC096030.1 | - | 2 | 495 | 30610161 | ||
BC118038.1 | - | 2 | 367 | 40093984 |