Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178641.5 | NP_848756.2 | 3 | 473 | ||
GenBank mRNA | AK030870.1 | - | 3 | 475 | ||
AK129249.1 | - | 3 | 430 | |||
AK150418.1 | - | 3 | 513 | |||
AK154996.1 | - | 3 | 478 | |||
AK211511.1 | - | 2 | 149 | |||
AK218585.1 | - | 2 | 148 | |||
BC125437.1 | - | 3 | 361 | 40129952 | ||
BC137700.1 | - | 3 | 361 | 8860719 |