Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003722.1 | NP_001003722.1 | 1, 2 | 221 | ||
NM_001499.2 | NP_001490.1 | 1, 2 | 221 | |||
XM_011518551.1 | XP_011516853.1 | 1, 2 | 221 | |||
GenBank mRNA | AF058922.1 | - | 1, 2 | 194 | ||
AK222842.1 | - | 1, 2 | 167 | |||
AK222854.1 | - | 1, 2 | 195 | |||
AK290533.1 | - | 1, 2 | 107 | |||
BC030012.1 | - | 1, 2 | 214 | 4829102 | ||
BM786801.1 | - | 1, 2 | 221 |