Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178676.4 | NP_848791.2 | 4 | 336 | ||
GenBank mRNA | AK032057.1 | - | 4 | 323 | ||
AK046218.1 | - | 4 | 312 | |||
AK087561.1 | - | 4 | 310 | |||
AY376710.1 | - | 4 | 304 | |||
AY714060.1 | - | 4 | 307 | |||
BC079871.1 | - | 4 | 307 | 6831925 |