Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164562.1 | NP_001158034.1 | 4 | 609 | ||
NM_178677.4 | NP_848792.2 | 5 | 734 | |||
GenBank mRNA | AK029985.1 | - | 5 | 665 | ||
AK031125.1 | - | 4 | 606 | |||
AK044249.1 | - | 4 | 609 | |||
BC046289.1 | - | 4 | 564 | 6469221 |