Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178679.2 | NP_848794.1 | 2 | 520 | ||
GenBank mRNA | AF543761.1 | - | 1 | 241 | ||
AJ516033.1 | - | 2 | 382 | |||
AK031846.1 | - | 2 | 316 | |||
AK033892.1 | - | 2 | 282 | |||
AK044322.1 | - | 2 | 279 | |||
AK077316.1 | - | 2 | 293 | |||
AK122387.1 | - | 2 | 507 | |||
BC080272.1 | - | 2 | 480 | 6837597 | ||
BC157983.1 | - | 2 | 321 | 9088140 | ||
BC157991.1 | - | 2 | 321 | 9088144 | ||
CB519185.1 | - | 2 | 520 |