Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178690.4 | NP_848805.2 | 3, 4 | 197 | ||
GenBank mRNA | AK076399.1 | - | 3, 4 | 180 | ||
AK080432.1 | - | 3, 4 | 179 | |||
AK134604.1 | - | 3, 4 | 179 | |||
AK164523.1 | - | 3, 4 | 178 | |||
AK170768.1 | - | 3, 4 | 169 | |||
AK172881.1 | - | 3, 4 | 184 | |||
BC046297.1 | - | 3, 4 | 163 | 6486170 | ||
CJ084350.1 | - | 3, 4 | 198 |