Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145876.2 | NP_001139348.1 | 2 | 890 | ||
NM_001145877.2 | NP_001139349.1 | 2 | 936 | |||
NM_001281795.1 | NP_001268724.1 | 2 | 811 | |||
NM_178696.5 | NP_848811.2 | 2 | 936 | |||
GenBank mRNA | AK032960.1 | - | 2 | 914 | ||
AK044648.1 | - | 2 | 783 | |||
AK046586.1 | - | 2 | 899 | |||
AK079532.1 | - | 2 | 898 | |||
AK083273.1 | - | 2 | 906 | |||
AK083470.1 | - | 2 | 786 | |||
AK134914.1 | - | 2 | 935 | |||
AK144731.1 | - | 2 | 863 | |||
AK165159.1 | - | 2 | 903 | |||
AK166011.1 | - | 2 | 908 | |||
AK166331.1 | - | 2 | 897 | |||
AK168955.1 | - | 2 | 909 | |||
AK169886.1 | - | 2 | 900 | |||
BC012234.1 | - | 2 | 715 | 4191504 | ||
BC052771.1 | - | 2 | 881 | 6310271 |