Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178697.4 | NP_848812.1 | 1, 2 | 271 | ||
GenBank mRNA | AK028704.1 | - | 1, 2 | 272 | ||
AK035512.1 | - | 1, 2 | 338 | |||
AK048276.1 | - | 1, 2 | 262 | |||
AY161007.1 | - | 1, 2 | 227 | |||
BC096379.1 | - | 1, 2 | 334 | 5317084 |