Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178720.4 | NP_848835.1 | 4 | 473 | ||
GenBank mRNA | AK034628.1 | - | 4 | 474 | ||
AK047981.1 | - | 3 | 383 | |||
AK052599.1 | - | 5 | 583 | |||
BC116254.1 | - | 4 | 442 | 40093631 | ||
BC116255.1 | - | 4 | 442 | 40093632 |