Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145977.1 | NP_001139449.1 | 3 | 903 | ||
NM_178721.4 | NP_848836.2 | 4 | 930 | |||
GenBank mRNA | AK033973.1 | - | 3 | 530 | ||
AK038842.1 | - | 4 | 909 | |||
AK043760.1 | - | 4 | 913 | |||
AK046800.1 | - | 3 | 806 | |||
AK163636.1 | - | 4 | 907 | |||
AK165030.1 | - | 4 | 927 | |||
BC139401.1 | - | 4 | 499 | 8862423 | ||
BC139402.1 | - | 4 | 499 | 8862424 |