Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178624.6 | NP_848739.1 | 15 | 2025 | ||
GenBank mRNA | AK013142.1 | - | 16 | 2161 | ||
AK039010.1 | - | 15 | 2025 | |||
AK044693.1 | - | 15 | 2022 | |||
AK045742.1 | - | 15 | 2012 | |||
AK141177.1 | - | 15 | 2015 | |||
AK163472.1 | - | 15 | 2021 | |||
BC096582.1 | - | 15 | 2011 | 3963599 | ||
BC145665.1 | - | 15 | 2005 | 40130968 | ||
BC145998.1 | - | 15 | 2005 | 40131301 |