Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297436.1 | NP_001284365.1 | 5 | 1986 | ||
NM_001523.3 | NP_001514.2 | 5 | 1989 | |||
GenBank mRNA | BC035837.1 | - | 5 | 1968 | 5589083 | |
D84424.1 | - | 4 | 1973 | |||
U59269.1 | - | 5 | 1965 |