Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178628.5 | NP_848743.1 | 15 | 2308 | ||
GenBank mRNA | AK046919.1 | - | 14 | 2038 | ||
AK050339.1 | - | 15 | 2145 | |||
AK077004.1 | - | 3 | 1430 | |||
AK083918.1 | - | 1174 | ||||
AK140185.1 | - | 14 | 2050 | |||
AK147311.1 | - | 15 | 5572 | |||
BC050784.1 | - | 14 | 2037 | 6595181 | ||
BC057068.1 | - | 10 | 1868 | 5689726 |