Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178652.2 | NP_848767.1 | 6 | 680 | ||
GenBank mRNA | AK050321.1 | - | 6 | 527 | ||
AK075916.1 | - | 6 | 680 | |||
AK087935.1 | - | 6 | 525 | |||
BC051540.1 | - | 5 | 410 | |||
BC096051.1 | - | 6 | 464 |