Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGFAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297439.1 | NP_001284368.1 | 6, 7 | 792 | ||
NM_001528.3 | NP_001519.1 | 6, 7 | 792 | |||
GenBank mRNA | AK314099.1 | - | 6, 7 | 750 | ||
BC112190.1 | - | 6, 7 | 792 | 8327658 | ||
BC112192.1 | - | 6, 7 | 804 | 8327660 | ||
D14012.1 | - | 6, 7 | 726 |