Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080381.1 | NP_001073850.1 | 10 | 1034 | ||
NM_001286100.1 | NP_001273029.1 | 10 | 902 | |||
NM_001286101.1 | NP_001273030.1 | 11 | 1127 | |||
NM_029679.2 | NP_083955.1 | 10 | 1034 | |||
NM_178658.5 | NP_848773.3 | 10 | 887 | |||
GenBank mRNA | AK031928.1 | - | 10 | 987 | ||
AK036452.1 | - | 11 | 1107 | |||
AK039271.1 | - | 10 | 999 | |||
AK040887.1 | - | 10 | 867 | |||
AK041665.1 | - | 10 | 888 | |||
AK084589.1 | - | 2 | 1759 | |||
AK122269.1 | - | 10 | 966 | |||
AK134538.1 | - | 10 | 965 | |||
AK163880.1 | - | 10 | 970 | |||
BC059179.1 | - | 10 | 902 |