Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB509340.1 | - | 2 | 229 | ||
AK040594.1 | - | 2 | 408 | |||
AK041386.1 | - | 2 | 445 | |||
AK041562.1 | - | 2 | 334 | |||
AK041622.1 | - | 2 | 408 | |||
AK088136.1 | - | 2 | 410 | |||
AK089077.1 | - | 2 | 410 | |||
AK138701.1 | - | 2 | 408 | |||
AK169558.1 | - | 2 | 178 | |||
BB638055.1 | - | 2 | 445 | |||
BC094898.1 | - | 171 | ||||
BC125486.1 | - | 2 | 291 | 40130001 | ||
BC125488.1 | - | 2 | 291 | 40130003 |