Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178675.4 | NP_848790.2 | 8 | 2060 | ||
GenBank mRNA | AK038601.1 | - | 8 | 2055 | ||
AK048957.1 | - | 8 | 2020 | |||
AK053426.1 | - | 8 | 2029 | |||
AK053545.1 | - | 8 | 1999 | |||
AK083768.1 | - | 8 | 1999 | |||
AK139331.1 | - | 8 | 2030 | |||
AK163806.1 | - | 8 | 1860 | |||
AK165021.1 | - | 8 | 2014 | |||
AK166268.1 | - | 8 | 1988 | |||
BC059075.1 | - | 8 | 1956 | 6826829 |