Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178676.4 | NP_848791.2 | 11 | 3020 | ||
GenBank mRNA | AK032057.1 | - | 11 | 3007 | ||
AK046218.1 | - | 11 | 2996 |