Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178679.2 | NP_848794.1 | 3 | 1132 | ||
GenBank mRNA | AF543761.1 | - | 2 | 853 | ||
AJ516033.1 | - | 3 | 994 | |||
AK031846.1 | - | 3 | 928 | |||
AK033892.1 | - | 3 | 894 | |||
AK044322.1 | - | 3 | 891 | |||
AK044655.1 | - | 2 | 215 | |||
AK077316.1 | - | 3 | 905 | |||
AK122387.1 | - | 3 | 1119 | |||
BC080272.1 | - | 3 | 1092 | 6837597 | ||
BC157983.1 | - | 3 | 933 | 9088140 | ||
BC157991.1 | - | 3 | 933 | 9088144 |