Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005335.1 | NP_001005335.1 | 4 | 468 | ||
NM_001533.2 | NP_001524.2 | 4 | 702 | |||
XM_011526886.1 | XP_011525188.1 | 4 | 491 | |||
XM_011526887.1 | XP_011525189.1 | 4 | 535 | |||
XM_011526889.1 | XP_011525191.1 | 4 | 586 | |||
GenBank mRNA | AB044547.1 | - | 4 | 697 | ||
AK292115.1 | - | 4 | 467 | |||
AK298384.1 | - | 4 | 538 | |||
AK301062.1 | - | 4 | 476 | |||
AK313652.1 | - | 4 | 637 | |||
BC069184.1 | - | 4 | 641 | |||
BI256815.1 | - | 4 | 468 | |||
X16135.1 | - | 4 | 626 |