Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178695.5 | NP_848810.3 | 6 | 1130 | ||
GenBank mRNA | AK035530.1 | - | 6 | 956 | ||
AK037087.1 | - | 6 | 1051 | |||
AK038703.1 | - | 6 | 1131 | |||
AK166405.1 | - | 5 | 1091 | |||
BC099390.1 | - | 6 | 1047 | 30715383 | ||
BC151011.1 | - | 6 | 1060 | 9087924 | ||
BC151019.1 | - | 6 | 1059 | 9087932 |