Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098476.2 | NP_001091946.1 | 6 | 811 | ||
NM_178700.5 | NP_848815.2 | 6 | 1139 | |||
GenBank mRNA | AK044889.1 | - | 6 | 1140 | ||
AK045986.1 | - | 6 | 1093 | |||
AK077782.1 | - | 6 | 1140 | |||
AK089014.1 | - | 5 | 675 | |||
AK168385.1 | - | 6 | 782 | |||
AK168459.1 | - | 6 | 782 | |||
AK210897.1 | - | 3 | 326 | |||
AK218546.1 | - | 3 | 326 |