Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178705.5 | NP_848820.4 | 4, 5 | 1049 | ||
GenBank mRNA | AK032204.1 | - | 4, 5 | 706 | ||
AK034077.1 | - | 4, 5 | 667 | |||
AK034240.1 | - | 4, 5 | 701 | |||
AK082126.1 | - | 4, 5 | 797 | |||
AY164459.1 | - | 4, 5 | 1046 | |||
BC075646.1 | - | 4, 5 | 679 | 30620475 |