Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178719.5 | NP_848834.2 | 6 | 2358 | ||
GenBank mRNA | AK038466.1 | - | 6 | 2330 | ||
AK041184.1 | - | 6 | 2251 | |||
AK050546.1 | - | 6 | 2318 | |||
AK080375.1 | - | 6 | 2305 | |||
BC094597.1 | - | 6 | 2290 | |||
BC137633.1 | - | 5 | 2258 | 8860652 |