Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1BG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130786.3 | NP_570602.2 | 3 | 232 | ||
GenBank mRNA | AB073611.1 | - | 3 | 225 | ||
AF414429.1 | - | 3 | 212 | |||
AK055885.1 | - | 3 | 225 | |||
AK289417.1 | - | 3 | 196 | |||
BC035719.1 | - | 3 | 279 | 5588885 | ||
BX325198.2 | - | 3 | 232 | |||
EU794585.1 | - | 3 | 224 |