Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001606.4 | NP_001597.2 | 32 | 5267 | ||
NM_212533.2 | NP_997698.1 | 32 | 5259 | |||
XM_006716996.3 | XP_006717059.1 | 31 | 5201 | |||
GenBank mRNA | AB028985.2 | - | 18 | 3124 | ||
AB177854.1 | - | 19 | 3168 | |||
AF178941.1 | - | 31 | 5169 | |||
AF327657.1 | - | 31 | 5125 | |||
AL833967.1 | - | 19 | 3207 | |||
BC008755.1 | - | 3 | 414 | |||
BC144588.1 | - | 31 | 5249 | |||
BF983101.1 | - | 5 | 511 |