Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271696.1 | NP_001258625.1 | 12 | 1656 | ||
NM_001271697.1 | NP_001258626.1 | 11 | 1536 | |||
NM_001271698.1 | NP_001258627.1 | 11 | 1578 | |||
NM_001271699.1 | NP_001258628.1 | 11 | 1539 | |||
NM_004299.4 | NP_004290.2 | 12 | 1659 | |||
GenBank mRNA | AF038950.1 | - | 12 | 1596 | ||
AF078777.1 | - | 12 | 1576 | |||
AF133659.1 | - | 12 | 1588 | |||
AK001418.1 | - | 12 | 1616 | |||
AK294657.1 | - | 11 | 1520 | |||
AK307414.1 | - | 11 | 1459 | |||
BC006323.2 | - | 12 | 1592 | 4138236 | ||
BT009918.1 | - | 12 | 1591 | |||
BX537833.1 | - | 11 | 1476 | |||
KU177872.1 | - | 12 | 1591 | |||
U66679.1 | - | 3 | 309 |