Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025091.1 | NP_001020262.1 | 18 | 1848 | ||
NM_001090.2 | NP_001081.1 | 17 | 1734 | |||
GenBank mRNA | AB209564.1 | - | 10 | 1011 | ||
AF027302.1 | - | 17 | 1733 | |||
AL832430.1 | - | 12 | 1278 | |||
BC034488.2 | - | 18 | 1793 | 4826422 | ||
BC112923.1 | - | 18 | 1781 | 7470309 | ||
U66677.1 | - | 8 | 809 |