Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000017.3 | NP_000008.1 | 2 | 296 | ||
NM_001302554.1 | NP_001289483.1 | 2 | 296 | |||
GenBank mRNA | AB527081.1 | - | 2 | 148 | ||
AK300645.1 | - | 2 | 174 | |||
BC025963.1 | - | 2 | 195 | 4842286 | ||
DB487184.1 | - | 2 | 299 | |||
DB505221.1 | - | 2 | 299 | |||
M26393.1 | - | 2 | 180 |